missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD9 Rat anti-Mouse, Clone: 479608, R&D Systems™
Rat Monoclonal Antibody
POA
Specifications
Antigen | CD9 |
---|---|
Clone | 479608 |
Immunogen | NS0 mouse myeloma cell line transfected with mouse CD9 Pro2-Val226 Accession # P40240 |
Host Species | Rat |
Primary or Secondary | Primary |
Description
CD9 Monoclonal specifically detects CD9 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, CyTOF-ready.Specifications
CD9 | |
NS0 mouse myeloma cell line transfected with mouse CD9 Pro2-Val226 Accession # P40240 | |
Primary | |
Mouse | |
Unconjugated | |
928 | |
Purified | |
Flow Cytometry, Immunohistochemistry, CyTOF | |
RUO |
479608 | |
Rat | |
Monoclonal | |
IgG2a | |
BA2, BA-2/p24 antigen, BTCC-1, CD9, CD9 antigen, CD9 molecule, Cell growth-inhibiting gene 2 protein, DRAP-27, Leukocyte antigen MIC3, MIC3, motility related protein-1,5H9 antigen, Motility-related protein, MRP-1FLJ99568, P24, Tetraspanin-29, TSPAN29, TSPAN-29, TSPAN29MIC3CD9 antigen (p24) | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only