missing translation for 'onlineSavingsMsg'
Learn More

CD8b Rat anti-Mouse, eFluor 450, Clone: eBioH35-17.2 (H35-17.2), eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 48-0083-82

UNSPSC : 12352200

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15301690

  • £167.38 / 100µg


EU Stock 3
Add to basket

Disclaimers:

For Research Use Only.


Specifications

Description

The eBioH35-17.2 monoclonal antibody reacts with the mouse CD8 beta molecule. The CD8 beta chain associates with the CD8 alpha chain to form the CD8 alpha/beta heterodimer expressed on the surface of a majority of thymocytes, and on peripheral cytotoxic alpha beta TCR T cells. CD8 binds to MHC class I and plays a role in T cell development and activation of mature T cells.
Description & Specifications

Specifications

CD8b
eBioH35-17.2 (H35-17.2)
eFluor 450
Conjugated
Rat
100μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Flow Cytometry
0.2mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
CD8 beta, Ly-3, Ly-C, Lyt-3
IgG2b
RUO
Primary
Murine