missing translation for 'onlineSavingsMsg'
Learn More

CD8b, Alexa Fluor 488, clone: eBioH35-17.2 (H35-17.2), eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 53-0083-81

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15341980

  • £78.00 / 50µg


Estimated Shipment date
from Supplier 30-01-2019
Add to basket

Description

Description

Description: The eBioH35-17.2 monoclonal antibody reacts with the mouse CD8 beta molecule. The CD8 beta chain associates with the CD8 alpha chain to form the CD8 alpha/beta heterodimer expressed on the surface of a majority of thymocytes, and on peripheral cytotoxic alpha beta TCR T cells. CD8 binds to MHC class I and plays a role in T cell development and activation of mature T cells.Applications Reported: This eBioH35-17.2 (H35-17.2) antibody has been reported for use in flow cytometric analysis.Applications Tested: This eBioH35-17.2 (H35-17.2) antibody has been tested by flow cytometric analysis of mouse splenocytes and thymocytes. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Excitation: 488 nm; Emission: 519 nm; Laser: Blue Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD8b
Monoclonal
0.5mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
P10300
Cd8b1
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Murine
Flow Cytometry
eBioH35-17.2 (H35-17.2)
Alexa Fluor 488
Cd8b1
Liquid
Cd8b1
IgG2b, kappa, kappa
50μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
12526
Videos

For Research Use Only.