missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD8a Rat, PE-Cy5, Clone: 53-6.7, enQuireBio™
Rat Monoclonal Antibody
£71.60 - £173.00
Specifications
Antigen | CD8a |
---|---|
Clone | 53-6.7 |
Concentration | 0.2mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15941568
|
EnquireBio
QAB16-PE5-25UG |
25 μg |
£71.60
25µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15931568
|
EnquireBio
QAB16-PE5-100UG |
100 μg |
£173.00
100µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
CD8a | |
0.2mg/mL | |
Monoclonal | |
Rat | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
12525 | |
IgG2a κ | |
Protein A/G Purified |
53-6.7 | |
Flow Cytometry | |
PE-Cyanine5 | |
RUO | |
NP_033987 | |
Cd8a | |
Primary | |
Store at 2-4°C. |