missing translation for 'onlineSavingsMsg'
Learn More

CD8a, PE-Cyanine5.5, clone: 53-6.7, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 35-0081-82

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15351020

  • £189.69 / 100µg


EU Stock 4
Add to basket

Description

Description

Description: The 53-6.7 monoclonal antibody reacts with the mouse CD8a molecule. CD8a is an approximately 32-34kDa cell surface receptor expressed either as a heterodimer with the CD8 beta chain (CD8 alpha beta) or as a homodimer (CD8 alpha alpha). A majority of thymocytes and a subpopulation of mature alpha beta TCR T cells express CD8 alpha beta while gamma delta TCR T cells, a subpopulation of intestinal intraepithelial lymphocytes (IELs) and dendritic cells express CD8 alpha alpha. CD8 binds to MHC class I and through its association with protein tyrosine kinase p56lck plays a role in T cell development and activation of mature T cells.Applications Reported: This 53-6.7 antibody has been reported for use in flow cytometric analysis.Applications Tested: This 53-6.7 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.125 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Light sensitivity: This tandem dye is sensitive photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 μL cell sample + 100 μL IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically.Excitation: 488-567 nm; Emission: 690 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser
Specifications

Specifications

CD8a
Monoclonal
0.2mg/mL
PBS with 0.09% sodium azide; pH 7.2
P01731
CD8, Leu-2, MAL, p32, CD8a
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Murine
Flow Cytometry
53-6.7
PE-Cyanine5.5
Cd8a
Conjugated
Cd8a
IgG2a, kappa
100μg
Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage, handling and experimental procedures.
Primary
12525
Videos

For Research Use Only.