missing translation for 'onlineSavingsMsg'
Learn More

CD8a Mouse, V450, Clone: Hit8a, enQuireBio™

Mouse Monoclonal Antibody

£109.00 - £219.00

Specifications

Antigen CD8a
Clone Hit8a
Concentration 5μL/Test
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15951658
View Documents Promotion Details
EnquireBio
QAB20-V450-25TESTS
25 Tests
£109.00
25 tests
View alternative products
Save Up to 
»
Estimated Shipment: 03-06-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15941658
View Documents Promotion Details
EnquireBio
QAB20-V450-100TESTS
100 Tests
£219.00
100 tests
View alternative products
Save Up to 
»
Estimated Shipment: 03-06-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

CD8a
5μL/Test
Monoclonal
Mouse
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2
925
IgG1 κ
Protein A/G Purified
Hit8a
Flow Cytometry
Violet Fluor 450 (V450)
RUO
NP_741969
CD8A
Primary
Store at 2-4°C.
Videos
SDS
Documents

Documents

Product Certifications