missing translation for 'onlineSavingsMsg'
Learn More

CD8a Mouse, PE-Cy7, Clone: RPA-T8, enQuireBio™

Mouse Monoclonal Antibody

Brand:  EnquireBio QAB19-PE7-100Tests

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15961618

  • £220.56 / 100 tests
Estimated Shipment: 17-01-2022

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

CD8a
RPA-T8
PE-Cyanine7
NP_741969
Mouse
Protein A/G Purified
RUO
Primary
925
Flow Cytometry
5μL/Test
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2
CD8A
IgG1 kappa
100 Tests
Store at 2-4°C.
Monoclonal
Videos
SDS
Documents
Certificates
Special Offers

Special Offers