missing translation for 'onlineSavingsMsg'
Learn More

CD8a, FITC, clone: OX8, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 11-0084-82

Code : NEW

Additional Details : Weight : 0.09500kg

 View more versions of this product

Product Code. 15516326

  • £93.40 / 100µg


EU Stock 1
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: The OX8 monoclonal antibody reacts with rat CD8 alpha. CD8 alpha is a member of the immunoglobulin superfamily which heterodimerizes with CD8 beta to form the CD8 antigen expressed on CD4+CD8+, double-positive thymocytes, the majority of MHC class I-restricted peripheral T cells, and on some macrophages. CD8 binds to MHC class I expressed on the surface of antigen-presenting cells during antigen presentation, and participates in T-cell receptor signal transduction through association with the kinase Lck.Applications Reported: This OX8 antibody has been reported for use in flow cytometric analysis.Applications Tested: This OX8 antibody has been tested by flow cytometric analysis of rat splenocytes. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD8a
OX8
FITC
Conjugated
Murine
100μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Flow Cytometry
0.5mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
CD8 alpha
IgG1
RUO
Primary
Rat
Videos