missing translation for 'onlineSavingsMsg'
Learn More

CD8a Mouse anti-Human, PE, Clone: SK1, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 12-0087-42

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15586696

  • £219.00 / 100 tests

In stock, ready to ship: 3

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Description: The SK1 monoclonal antibody reacts with the human CD8a molecule, an approximately 32-34 kDa cell surface receptor expressed either as a heterodimer with the CD8 beta chain (CD8 alpha/beta) or as a homodimer (CD8 alpha/alpha). A majority of thymocytes and a subpopulation of mature T cells and NK cells express CD8a. CD8 binds to MHC class I and through its association with protein tyrosine kinase p56lck plays a role in T-cell development and activation of mature T cells. Applications Reported: This SK1 (SK-1) antibody has been reported for use in flow cytometric analysis. Applications Tested: This SK1 (SK-1) antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 µL (0.125 µg) per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered.

CD8 (Cluster of Differentiation 8) is a cell surface glycoprotein found on most cytotoxic T lymphocytes that mediate efficient cell-cell interactions within the immune system. The CD8 antigen acts as a co-receptor with the T-cell receptor on the T lymphocyte to recognize antigens displayed by an antigen presenting cell (APC) in the context of class I MHC molecules. CD8 is found on a T cell subset of normal cytotoxic/suppressor cells which make up approximately 20-35 % of human peripheral blood lymphocytes. The CD8 antigen is also detected on natural killer (NK) cells, subpopulations of peripheral blood null cells, thymocytes and bone marrow cells. The CD8 co-receptor functions as either a homodimer composed of two alpha chains, or as a disulfide-linked heterodimer composed of one alpha and one beta chain. Both alpha and beta chains share significant homology to immunoglobulin variable light chains. The majority of CD8+ T cells express CD8 as a alpha/beta heterodimer. In HIV, the HIV-2 envelope glycoprotein binds CD8 alpha chain (but not the beta chain).
Specifications

Specifications

CD8a
Monoclonal
5 μL/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
P01732
CD8, Leu-2, MAL, p32, CD8a
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
SK1
PE
CD8A
Liquid
CD8A
IgG1, kappa
100 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
925
Videos
SDS
Documents

For Research Use Only.