missing translation for 'onlineSavingsMsg'
Learn More

CD8a Mouse anti-Human, Biotin, Clone: AMC908, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 13-0008-82

Code : NEW

Additional Details : Weight : 0.09500kg

Product Code. 15577306

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Description

Description

This AMC908 monoclonal antibody reacts with the human CD8a molecule, an approximately 32-34kDa cell surface receptor expressed either as a heterodimer with the CD8 β chain (CD8 αβ) or as a homodimer (CD8 αα). A majority of thymocytes and a subpopulation of mature T cells and NK cells express CD8a. CD8 binds to MHC class I and through its association with protein tyrosine kinase p56lck plays a role in T-cell development and activation of mature T cells.
Specifications

Specifications

CD8a
AMC908
Biotin
Conjugated
Murine
100μg
Store at 2-8°C. Do not freeze.
Monoclonal
Immunohistochemistry, Western Blot
0.5mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
CD8 alpha, leu-2a
IgG2a, kappa
RUO
Primary
Human
Videos
Documents

For Research Use Only.