missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD8A (Cytotoxic-&Suppressor T-Cell Marker) Mouse, Clone: rC8/468, enQuireBio™
Mouse Monoclonal Antibody
£181.83 - £395.00
Specifications
Clone | rC8/468 |
---|---|
Concentration | 200μg/mL |
Applications | Flow Cytometry, Immunofluorescence, Immunohistochemistry |
Classification | Monoclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15951178
|
EnquireBio
925-MSM11-P1 |
100 μg |
£395.00
100µg |
Estimated Shipment: 12-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15941178
|
EnquireBio
925-MSM11-P0 |
20 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
rC8/468 | |
Flow Cytometry, Immunofluorescence, Immunohistochemistry | |
Unconjugated | |
RUO | |
925 | |
IgG1 | |
Protein A/G purified from Bioreactor concentrate. | |
Cell Surface | |
HuT78 or hPBL. Tonsil |
200μg/mL | |
Monoclonal | |
Mouse | |
10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide. | |
Recombinant full-length human CD8 protein | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. | |
32kDa |