missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD8A (Cytotoxic-&Suppressor T-Cell Marker) Mouse, Clone: rC8/468, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 925-MSM11-P0
Additional Details : Weight : 0.01000kg
Specifications
CD8A (Cytotoxic-&Suppressor T-Cell Marker) | |
Cell Surface | |
rC8/468 | |
Unconjugated | |
Mouse | |
32kDa | |
Protein A/G purified from Bioreactor concentrate. | |
RUO | |
925 | |
IgG1 |
Flow Cytometry, Immunofluorescence, Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide. | |
Recombinant full-length human CD8 protein | |
HuT78 or hPBL. Tonsil | |
20 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |