missing translation for 'onlineSavingsMsg'
Learn More

CD87 (UPAR), PerCP-eFluor™ 710, clone: VIM5, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 46-3879-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15569246

  • £113.24 / 25 tests

Estimated Shipment date
from Supplier 27-04-2018
Add to basket



Description: This VIM5 monoclonal antibody reacts with human CD87, a GPI-linked receptor for urokinase plasminogen activator (uPA). By sequestering uPA, CD87 is involved in the conversion of plasminogen to plasmin on the leading edge of migrating cells, a process critical for chemotaxis. CD87 is expressed on the surface of monocytes, dendritic cells, and on activated T and NK cells. Interestingly, in granulocytes, CD87 expressed in the cytoplasm and on the cell surface. Non-hematopoietic cells such as endothelial cells, keratinocytes, and fibroblasts, also express this receptor. Finally, CD87 plays a role in cell adhesion and migration, angiogenesis, and tumor metastasis.This monoclonal antibody has been reported to have neutralization activity.Applications Reported: This VIM5 antibody has been reported for use in flow cytometric analysis.Applications Tested: This VIM5 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. PerCP-eFluor® 710 emits at 710 nm and is excited with the blue laser (488 nm); it can be used in place of PerCP-Cyanine5.5. We recommend using a 710/50 bandpass filter, however, the 695/40 bandpass filter is an acceptable alternative. Please make sure that your instrument is capable of detecting this fluorochrome.Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light.Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 μL of cell sample + 100 μL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically.Excitation: 488 nm; Emission: 710 nm; Laser: Blue Laser.Filtration: 0.2μm post-manufacturing filtered.


PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2
PLAUR, U-PAR, UPAR, URKR, monocyte activation antigen Mo3, u-plasminogen activator receptor form 2, urokinase plasminogen activator surface receptor
Affinity chromatography
Flow Cytometry
PerCP-eFluor 710
IgG1, kappa
25 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage, handling and experimental procedures.

For Research Use Only.