missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD86 Rat, PE, Clone: PO3.1, enQuireBio™
Rat Monoclonal Antibody
Brand: EnquireBio QAB93-PE-100ug
Additional Details : Weight : 0.01000kg
Specifications
CD86 | |
Monoclonal | |
0.2mg/mL | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
Cd86 | |
Protein A/G Purified | |
RUO | |
12524 | |
IgG2b κ |
Flow Cytometry | |
PO3.1 | |
PE | |
NP_062261 | |
Rat | |
100 μg | |
Primary | |
Store at 2-4°C. |