missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD86 Rat, PE, Clone: GL-1 (GL1), enQuireBio™
Rat Monoclonal Antibody
£71.60 - £182.00
Specifications
Antigen | CD86 |
---|---|
Clone | GL-1 (GL1) |
Concentration | 0.5mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15962158
|
EnquireBio
QAB51-PE-25UG |
25 μg |
£71.60
25µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15952158
|
EnquireBio
QAB51-PE-100UG |
100 μg |
£182.00
100µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
CD86 | |
0.5mg/mL | |
Monoclonal | |
Rat | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
12524 | |
IgG2a κ | |
Protein A/G Purified |
GL-1 (GL1) | |
Flow Cytometry | |
PE | |
RUO | |
NP_062261 | |
Cd86 | |
Primary | |
Store at 2-4°C. |