Learn More
CD85k (ILT3) Monoclonal Antibody (ZM4.1), APC, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
£105.00 - £242.00
Specifications
Antigen | CD85k (ILT3) |
---|---|
Clone | ZM4.1 |
Concentration | 5 μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15528116
|
Affymetrix eBioscience
17-5139-42 |
100 Tests |
£242.00
100 tests |
Estimated Shipment: 30-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15518116
|
Affymetrix eBioscience
17-5139-41 |
25 Tests |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Description: The monoclonal antibody ZM4.1 recognizes ILT3, also known as CD85k. ILT3 is a member of the large family of ILT molecules (Immunologulin-like Transcripts, Leucocyte Ig-like Receptors (LIRs) and Monocyte/Macrophage Ig-like Receptors (MIRs). ILT3 is a 60kDa transmembrane protein with two Ig domains and ITIMs. Expression is found on monocytes, macrophages, and dendritic cells. High levels of ILT3 expression is also found on plasmacytoid dendritic cells (pDCs). ILT3 expression by DCs has been shown to be required to induce Regulatory Tcells (tregs). Addition of ZM4.1, like ZM3.8, blocks the suppressive function of the Tregs in in vitro culture systems with allogenic DC's. Applications Reported: This ZM4.1 antibody has been reported for use in flow cytometric analysis. Applications Tested: This ZM4.1 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.125 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser. Filtration: 0.2 μm post-manufacturing filtered.
CD85k (ILT3) Monoclonal antibody specifically detects CD85k (ILT3) in Human samples. It is validated for Flow Cytometry.Specifications
CD85k (ILT3) | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
Q8NHJ6 | |
LILRB4 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
LILRB4 |
ZM4.1 | |
Flow Cytometry | |
APC | |
Mouse | |
Human | |
11006 | |
IgG1 κ | |
Affinity chromatography | |
Antibody |
For Research Use Only.