missing translation for 'onlineSavingsMsg'
Learn More

CD85j (ILT2) Monoclonal Antibody (HP-F1), APC, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 17-5129-41

76.67 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.



Code : NEW

Additional Details : Weight : 0.23750kg

Product Code. 15598106

  • £115.00 / 25 tests
Estimated Shipment: 08-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

CD85j (ILT2) Monoclonal antibody specifically detects CD85j (ILT2) in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

CD85j (ILT2)
Monoclonal
5 μL/Test
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2
Q8NHL6
LILRB1
Affinity chromatography
RUO
10859
4° C, store in dark, DO NOT FREEZE!
IgG1 κ
Flow Cytometry
HP-F1
APC
LILRB1
CD85 antigen-like family member J; CD85j; Ig-like transcript 2; ILT2; ILT-2; Immunoglobulin-like transcript 2; leucocyte Ig-like receptor B1; leukocyte immunoglobulin like receptor B1; leukocyte immunoglobulin-like receptor subfamily B member 1; leukocyte immunoglobulin-like receptor subfamily B member 1 soluble isoform; leukocyte immunoglobulin-like receptor subfamily B member 3-like; leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), member 1; LILRB1; LIR1; LIR-1; LOC100912456; MIR7; MIR-7; Monocyte/macrophage immunoglobulin-like receptor 7; myeloid inhibitory receptor 7; PIRB; PIR-B
Mouse
25 Tests
Primary
Human
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only.