missing translation for 'onlineSavingsMsg'
Learn More

CD85g (ILT7) Mouse anti-Human, PE, Clone: eBio17G10.2 (17G10.2), eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 12-5179-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15596996

  • £91.75 / 25 tests

Estimated Shipment
30-11-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes an immunoglobulin-like cell surface protein preferentially expressed in plasmacytoid dendritic cells (PDCs). This gene is highly expressed in PDCs, and is found to be rapidly down-regulated by interleukin 3 (IL3). This gene is one of the 19 highly related genes that form a leukocyte receptor gene cluster (LRC) at chromosomal region 19q13.4.
Specifications

Specifications

CD85g (ILT7)
Monoclonal
5 μL/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
P59901
CD85g, ILT7, CD85 antigen-like family member G, ILT-7, immunoglobulin-like transcript 7, leukocyte immunoglobulin-like receptor subfamily A member 4, leukocyte immunoglobulin-like receptor, subfamily A (without TM domain), member 4
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
eBio17G10.2 (17G10.2)
PE
LILRA4
Liquid
LILRA4
IgG1
25 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
23547
Videos
Documents

For Research Use Only.