missing translation for 'onlineSavingsMsg'
Learn More

CD83, Biotin, clone: Michel-17 (Michel17), eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 13-0831-80

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15279629

  • £56.31 / 25µg


Estimated Shipment date
from Supplier 04-06-2018
Add to basket

Description

Description

Description: The Michel-17 monoclonal antibody reacts with mouse CD83, a 45kDa cell surface glycoprotein and a member of the Ig superfamily. The mouse CD83 antigen is expressed predominantly on mature DC and activated lymphocytes. Cross-linking of CD83 with Michel-17 on DC or activated T cells does not induce any activation signal. CD83 plays an important role in T cell development through interaction with its ligand. CD83-Ig protein has revealed the presence of a CD83 ligand expressed mainly by B220^+ cells in mouse spleen.Applications Reported: This Michel-17 (Michel17) antibody has been reported for use in flow cytometric analysis.Applications Tested: This Michel-17 (Michel17) antibody has been tested by flow cytometric analysis of LPS-activated mouse splenocytes. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD83
Monoclonal
0.5mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
O88324
HB15
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Murine
Flow Cytometry
Michel-17 (Michel17)
Biotin
Cd83
Conjugated
Cd83
IgG1
25μg
Store at 2-8°C. Do not freeze.
Primary
12522
Videos

For Research Use Only.