Learn More
CD83 Monoclonal Antibody (HB15e), PE-Cyanine7, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
£92.25 - £265.00
Specifications
Antigen | CD83 |
---|---|
Clone | HB15e |
Concentration | 5 μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15578436
|
Affymetrix eBioscience
25-0839-42 |
100 Tests |
£265.00
100 tests |
Estimated Shipment: 28-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15568436
|
Affymetrix eBioscience
25-0839-41 |
25 Tests |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Description: The HB15e monoclonal antibody reacts with human CD83, a 45 kDa transmembrane glycoprotein. CD83, a member of the Ig superfamily, is expressed on cultured dendritic cells, interdigitating, follicular, and circulating dendritic cells as well as some proliferating lymphocytes, and human cell lines express this antigen. While the function of CD83 is unclear, it can serve as a useful marker for mature human blood dendritic cells. Applications Reported: This HB15e antibody has been reported for use in flow cytometric analysis. Applications Tested: This HB15e antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Light sensitivity: This tandem dye is sensitive photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 μL cell sample + 100 μL IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation.
CD83 Monoclonal antibody specifically detects CD83 in Human samples. It is validated for Flow Cytometry.Specifications
CD83 | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
Q01151 | |
CD83 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
CD83 |
HB15e | |
Flow Cytometry | |
PE-Cyanine7 | |
Mouse | |
Human | |
9308 | |
IgG1 κ | |
Affinity chromatography | |
Antibody |
For Research Use Only.