missing translation for 'onlineSavingsMsg'
Learn More

CD83 Mouse anti-Human, PE, Clone: HB15e, eBioscience™

Mouse Monoclonal Antibody

£85.00 - £184.00

Specifications

Antigen CD83
Clone HB15e
Target Species Human
Conjugate PE
Isotype IgG1, kappa
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15586826
View Documents
Affymetrix eBioscience
12-0839-41
25 Tests £85.00
25 tests View alternative products
Save Up to 
»
Estimated Shipment
28-10-2020
Please to purchase this item. Need a web account? Register with us today!
15596826
View Documents
Affymetrix eBioscience
12-0839-42
100 Tests £184.00
100 tests View alternative products
Save Up to 
»
In stock, ready to ship: 8 Please to purchase this item. Need a web account? Register with us today!
Description

Description

CD83 cell surface antigen is a 40-45kD glycoprotein expressed by peripheral blood dendritic cells. Peripheral lymphocytes can be induced to express very low levels of CD83 after culture in agents such as Con A or PHA. In immunohistology, CD83 is shown to be expressed strongly by interfollicular interdigitating reticulum cells and more weakly by cells within germinal centres. CD83 is also expressed by Langerhan′s cells in the skin. The CD83 antigen is a 186-amino-acid single-chain glycoprotein and this molecule is a member of the immunoglobulin superfamily that is composed of an extracellular V-type Ig-like single domain, a transmembrane region, and a short, 40-amino-acid cytoplasmic tail. CD83 antigen undergoes extensive post-translational glycosylation, since the determined Mr is twice the predicted size of the core protein. However, CD83+ cells have a unique cell surface immuno-phenotype that does not correlate with that of T cells, B cells, NK cells, or cells of the myelomonocytic lineage. CD83+ cells coexpress the highest levels of MHC class II molecules, when compared with other leucocyte lineages. They also co-express T cell markers (CD2, CD5), B cell markers (CD40, CD78), myeloid cell markers (CD13, CD33, CD36), cytokine receptors as well as other cell surface molecules. Diseases associated with CD83 dysfunction include plague and Rift Valley Fever.
Specifications

Specifications

CD83
Human
IgG1, kappa
Monoclonal
Liquid
Antibody
4° C, store in dark, DO NOT FREEZE!
HB15e
PE
Flow Cytometry
Primary
Monoclonal
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
RUO

For Research Use Only.