missing translation for 'onlineSavingsMsg'
Learn More

CD81, Biotin, clone: Eat-2 (EAT2), eBioscience™

Armenian Hamster Monoclonal Antibody

Brand:  Affymetrix eBioscience 13-0811-82

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15269629

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Description

Description

The Eat-2 (EAT2) monoclonal antibody reacts with mouse CD81, a molecule also known as TAPA-1. CD81 is expressed on CD4+CD8+ thymocytes and broadly in the periphery with high level expressed by B cells, NK cells, macrophages and dendritic cells and lower level on resting T cells. Activation of T cells induces upregulation of CD81. CD81 is involved in homotypic adhesion of B and T cells as well as costimulation of T cells. The Eat-2 antibody is reported to cross-react with rat CD81.
Specifications

Specifications

CD81
Eat-2 (EAT2)
Biotin
Conjugated
Armenian Hamster
100μg
Store at 2-8°C. Do not freeze.
Monoclonal
Flow Cytometry
0.5mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
TAPA-1, TAPA1
IgG
RUO
Primary
Murine
Videos

For Research Use Only.