missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD80 Monoclonal Antibody (1G10), APC, Invitrogen™
Rat Monoclonal Antibody
Brand: Invitrogen A14724
78.67 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.01000kg
Description
CD80 Monoclonal antibody specifically detects CD80 in Mouse samples. It is validated for Flow Cytometry.Specifications
CD80 | |
Monoclonal | |
0.1 mg/mL | |
PBS with <0.1% sodium azide | |
Q00609 | |
CD80 | |
Dibutyryl cAMP-Activated 5C2 cells | |
25 μg | |
Primary | |
Mouse | |
Antibody | |
IgG2a κ |
Flow Cytometry | |
1G10 | |
APC | |
CD80 | |
Activation B7-1 antigen; B7; B7 protein; B7.1; B71; B7-1; B7-1 protein; B7-1 protein precursor; BB1; B-lymphocyte activation antigen B7; Cd28l; CD28LG; CD28LG1; CD80; Cd80 a rat homolog of the human CD28/CTLA - 4 ligand (B7-1); CD80 anitgen; CD80 antigen; CD80 antigen (CD28 antigen ligand 1, B7-1 antigen); Cd80 molecule; CD80 protein precursor; costimulatory factor CD80; costimulatory molecule variant IgV-CD80; CTLA-4 counter-receptor B7.1; LAB7; Ly53; Ly-53; MIC17; sCD 80; sCD80; secreted B7-1 protein precursor; soluble CD 80; soluble CD80; T-cell costimulatory molecule; T-cell co-stimulatory protein B7-1; T-lymphocyte activation antigen CD80; TS/A-1; TSA1 | |
Rat | |
purified | |
RUO | |
12519 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |