missing translation for 'onlineSavingsMsg'
Learn More

CD80 (B7-1), FITC, clone: 2D10.4, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 11-0809-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15516456

  • £49.78 / 25 tests


EU Stock 3
Add to basket

Description

Description

Description: The 2D10.4 monoclonal antibody reacts with human CD80 (B7-1), a 60kDa member of the Ig superfamily. CD80 is expressed by activated B cells, macrophages and dendritic cells. In addition, activated T cells express this antigen. CD80 has high affinity for binding to two T cell surface antigens, CD28 and CD152 (CTLA-4). The interaction of CD28 and CD152 with CD80 is crucial in T-B cell communication leading to activation of T and B cells, respectively.Applications Reported: The 2D10.4 antibody has been reported for use in flow cytometric analysis.Applications Tested: This 2D10.4 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (1 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD80
Monoclonal
5μl/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
P33681
B7-1, B7.1, Cd28l, Ly-53, Ly53, MIC17, TS/A-1, LAB7, B7.1, CD28LG, CD28LG1
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
2D10.4
FITC
CD80
Conjugated
CD80
IgG1, kappa
25 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
941
Videos

For Research Use Only.