missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD80 Armenian Hamster, FITC, Clone: 16-10A1, enQuireBio™
Armenian Hamster Monoclonal Antibody
Brand: EnquireBio QAB50-F-500ug
Additional Details : Weight : 0.01000kg
Specifications
CD80 | |
Monoclonal | |
0.5mg/mL | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
Cd80 | |
Protein A/G Purified | |
RUO | |
12519 | |
IgG |
Flow Cytometry | |
16-10A1 | |
FITC | |
XP_017172347 | |
Armenian Hamster | |
500 μg | |
Primary | |
Store at 2-4°C. |