missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD8 Mouse, PE, Clone: SK1, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio QAB18-PE-100Tests
Additional Details : Weight : 0.01000kg
Specifications
CD8 | |
Monoclonal | |
5μL/Test | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
CD8A | |
Protein A/G Purified | |
RUO | |
925 | |
IgG1 κ |
Flow Cytometry | |
SK1 | |
PE | |
NP_741969 | |
Mouse | |
100 Tests | |
Primary | |
Store at 2-4°C. |