missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD8 Monoclonal Antibody (3B5), FITC, Invitrogen™
Mouse Monoclonal Antibody
£192.00 - £610.00
Specifications
Antigen | CD8 |
---|---|
Clone | 3B5 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | FITC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10130343
|
Invitrogen
MHCD0801 |
500 μL |
£192.00
Each |
Estimated Shipment: 16-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10661454
|
Invitrogen
MHCD08014 |
2 mL |
£610.00
Each |
Estimated Shipment: 16-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Commonly used, FITC conjugates provide relatively high absorptivity, excellent fluorescence quantum yield, and good water solubility.
CD8 Monoclonal antibody specifically detects CD8 in Human samples. It is validated for Flow Cytometry.Specifications
CD8 | |
Flow Cytometry | |
FITC | |
Mouse | |
Human | |
P01732, P10966 | |
925, 926 | |
Human CD8 | |
Primary | |
4° C, store in dark |
3B5 | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 4mg/mL BSA and 0.1% sodium azide | |
BB154331; CD8; CD8 alpha; CD8 alpha chain; CD8 alpha chain precursor; CD8 antigen; CD8 antigen 32 kDa chain; CD8 antigen 37 kDa chain; CD8 antigen alpha polypeptide; CD8 antigen alpha protein; CD8 antigen alpha protein precursor; CD8 antigen alpha-chain; CD8 antigen beta polypeptide; CD8 antigen beta polypeptide precursor; CD8 antigen beta-chain; CD8 antigen, alpha chain; CD8 antigen, alpha polypeptide; CD8 antigen, alpha polypeptide (p32); CD8 antigen, alpha-chain; CD8 antigen, beta chain; CD8 antigen, beta chain 1; CD8 antigen, beta polypeptide; CD8 antigen, beta polypeptide 1 (p37); CD8 antigen, beta-chain; CD8 beta; CD8 beta chain; CD8 beta-2; CD8a; CD8A antigen alpha; CD8a molecule; CD8A; T-cell surface glycoprotein; CD8alpha; CD8B; CD8b antigen; CD8b molecule; CD8b molecule pseudogene; Cd8b1; CD8beta; CD8BP; fCD8; LEU2; Leu-2; Leu2 T-lymphocyte antigen; leu-2a; Ly-2; LY3; Ly-3; Ly-35; Ly-B; Ly-C; Lymphocyte antigen 3; Lyt2; Lyt-2; Lyt-2.1 lymphocyte differentiation antigen (AA at | |
Cd8a, CD8B | |
IgG2a | |
purified | |
Antibody |