missing translation for 'onlineSavingsMsg'
Learn More

CD8a Monoclonal Antibody (OKT8 (OKT-8)), Alexa Fluor™ 700, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 56-0086-42

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15590327

  • £218.00 / 100 tests
In Stock
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Description: The OKT8 monoclonal antibody reacts with the human CD8a molecule, an approximately 32-34 kDa cell surface receptor expressed either as a heterodimer with the CD8 beta chain (CD8 alpha beta) or as a homodimer (CD8 alphaalpha). A majority of thymocytes and a subpopulation of mature T cells and NK cells express CD8a. CD8 binds to MHC class I and through its association with protein tyrosine kinase p56lck plays a role in T-cell development and activation of mature T cells. Preliminary testing indicates that OKT8 and two other mouse anti-human CD8 antibodies (clone RPA-T8, Cat. No.14-0088 and clone HIT8a, Cat. No.14-0089) do not compete with each other for binding to human peripheral blood leukocytes by flow cytometric analysis, suggesting that they do not bind to similar epitopes or block each other by steric hindrance. Applications Reported: This OKT8 (OKT-8) antibody has been reported for use in flow cytometric analysis. Applications Tested: This OKT8 (OKT-8) antibody has been tested by flow cytometric analysis of human peripheral blood leukocytes. This can be used at less than or equal to 0.125 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

CD8a Monoclonal antibody specifically detects CD8a in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

CD8a
Monoclonal
5 μL/Test
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2
P01732, P10966
Cd8a, CD8B
Affinity chromatography
RUO
925, 926
4° C, store in dark, DO NOT FREEZE!
Liquid
Flow Cytometry
OKT8 (OKT-8)
Alexa Fluor 700
Cd8a
BB154331; CD8; CD8 alpha; CD8 alpha chain; CD8 alpha chain precursor; CD8 antigen; CD8 antigen 32 kDa chain; CD8 antigen 37 kDa chain; CD8 antigen alpha polypeptide; CD8 antigen alpha protein; CD8 antigen alpha protein precursor; CD8 antigen alpha-chain; CD8 antigen beta polypeptide; CD8 antigen beta polypeptide precursor; CD8 antigen beta-chain; CD8 antigen, alpha chain; CD8 antigen, alpha polypeptide; CD8 antigen, alpha polypeptide (p32); CD8 antigen, alpha-chain; CD8 antigen, beta chain; CD8 antigen, beta chain 1; CD8 antigen, beta polypeptide; CD8 antigen, beta polypeptide 1 (p37); CD8 antigen, beta-chain; CD8 beta; CD8 beta chain; CD8 beta-2; CD8a; CD8A antigen alpha; CD8a molecule; CD8A; T-cell surface glycoprotein; CD8alpha; CD8B; CD8b antigen; CD8b molecule; CD8b molecule pseudogene; Cd8b1; CD8beta; CD8BP; fCD8; LEU2; Leu-2; Leu2 T-lymphocyte antigen; leu-2a; Ly-2; LY3; Ly-3; Ly-35; Ly-B; Ly-C; Lymphocyte antigen 3; Lyt2; Lyt-2; Lyt-2.1 lymphocyte differentiation antigen (AA at
Mouse
100 Tests
Primary
Human
Antibody
IgG2a κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only.