Learn More
CD79a Monoclonal Antibody (HM47), APC-eFluor™ 780, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Affymetrix eBioscience 47-0792-42
192.00 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.23750kg
Description
Description: The monoclonal antibody HM47 recognizes the cytoplasmic domain of CD79a, also known as mb-1. CD79a is a 47 kDa membrane glycoprotein that associates with CD79b to form the heterodimeric B cell receptor (BCR). This receptor is responsible for
CD79a Monoclonal antibody specifically detects CD79a in Baboon, Bovine, Canine, Chicken, Chimpanzee, Equine, Guinea Pig, Human, Mouse, Non-human Primate, Porcine, Rat, Rhesus Monkey samples. It is validated for Flow Cytometry.Specifications
CD79a | |
Monoclonal | |
5 μL/Test | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2 | |
P0CAN6, P11911, P11912, P40293 | |
CD79A | |
Affinity chromatography | |
RUO | |
100052219, 100190992, 100724815, 100913063, 12518, 281674, 484483, 722190, 740560, 973 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
HM47 | |
APC-eFluor 780 | |
CD79A | |
B-cell antigen receptor; B-cell antigen receptor complex-associated protein alpha chain; B-cell antigen receptor complex-associated protein alpha chain-like; CD79; CD79a; CD79A antigen (immunoglobulin-associated alpha); CD79a molecule; CD79a molecule, immunoglobulin-associated alpha; CD79A protein; CD79-alpha protein; EGK_10665; Ig alpha; IGA; Igalpha; ig-alpha; immunoglobulin-associated alpha; LOC100913063; Ly54; Ly-54; MB1; MB-1; MB-1 membrane glycoprotein; Membrane-bound immunoglobulin-associated protein; surface IgM-associated protein | |
Mouse | |
100 Tests | |
Primary | |
Baboon, Porcine, Rat, Rhesus Monkey, Bovine, Equine, Chicken, Human, Mouse, Non-human Primate, Human, Mouse, Non-human Primate | |
Antibody | |
IgG1 κ |
For Research Use Only.