missing translation for 'onlineSavingsMsg'
Learn More

CD73, eFluor 450, clone: eBioTY/11.8 (TY/11.8), eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 48-0731-80

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15391730

  • £70.03 / 25µg


EU Stock 4
Add to basket

Description

Description

Description: eBioTY/11.8 recognizes the with CD73 a 69-kDa GPI-anchored cell-surface protein with ecto-5'-nucleotidase activity. Expression on myeloid cells (CD11b) is restricted to the bone marrow. In human CD73 can be induced to secrete a soluble form with IL-2 suggesting a role in mediating activation signals. Differences between human and mouse CD73 have been reported. BALB/c mice have more CD4+CD73+ than CD8+CD73+ while the reciprocal is documented in humans.CD73 is expressed on a subset of lymphocytes and increases during lymphocyte maturation. Recently it has been found that memory CD4 T cells express and are similar to the uncommitted primed precursor helper cells (Thpp) that can differentiate into Th1 or Th2 cells. Furthermore CD73 has been found on regulatory T cells.Applications Reported: This eBioTY/11.8 (TY/11.8) antibody has been reported for use in flow cytometric analysis.Applications Tested: This eBioTY/11.8 (TY/11.8) antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.06 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.eFluor® 450 is an alternative to Pacific Blue®. eFluor® 450 emits at 445 nm and is excited with the Violet laser (405 nm). Please make sure that your instrument is capable of detecting this fluorochome.Excitation: 405 nm; Emission: 445 nm; Laser: Violet Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD73
Monoclonal
0.2mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
Q61503
CD73, E5NT, NT, NT5, NTE, eN, eNT, 5'-NT, 5'-nucleotidase, Purine 5-Prime-Nucleotidase, ecto-5'-nucleotidase
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Murine
Flow Cytometry
eBioTY/11.8 (TY/11.8)
eFluor 450
Nt5e
Conjugated
Nt5e
IgG1
25μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
23959
Videos

For Research Use Only.