CD73 Mouse anti-Human, PE-Cy7, Clone: AD2, eBioscience™

Mouse Monoclonal Antibody

Overview
Brand: Affymetrix eBioscience

Manufacturer Part Number: 25-0739-41

Code: NEW

Additional Details:
Additional Details: Weight: 0.23750kg



Disclaimers: For Research Use Only.

Product Code. 15528436

Quantity Price
1 £ 74.51 / 25 tests
Estimated Shipment date
from Supplier 05-06-2017
Add to basket


Description and Specification

Specification

Antigen CD73
Applications Flow Cytometry
Clone AD2
Concentration 5μL (0.25μg)/test
Conjugate PE-Cy7
Format Conjugated
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Gene Alias Ecto-5'-nucleotidase, NT5E
Host Species Murine
Isotype IgG1
Quantity 25 Tests
Regulatory Status RUO
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Target Species Human

This AD2 monoclonal antibody reacts with human CD73, a 5’-ectonucleotidase that converts 5’-adenosine monophosphate to adenosine. CD73 is expressed on the surface of endothelial cells, as well as B and T cells, including some CD4+Foxp3+ regulatory T cells. Adenosine production by these cells has been linked to the inhibition of CD4 T cell effector functions such as proliferation and cytokine secretion.