missing translation for 'onlineSavingsMsg'
Learn More

CD71 (Transferrin Receptor), FITC, clone: R17217 (RI7 217.1.4), eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 11-0711-81

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15268129

  • £56.56 / 50µg


EU Stock 1
Add to basket

Description

Description

Description: The R17217 monoclonal antibody reacts with mouse CD71, a 170-180kDa type II transmembrane protein. CD71, the transferrin receptor, exists as a homodimer on the cell surface and is essential for cellular growth. CD71 is expressed by immature proliferating cells and at low levels on resting mature lymphocytes. Antigen or mitogen stimulation of T and B cells upregulates the expression of CD71. Expression level differences have been observed in different mouse strains.Applications Reported: The R17217 antibody has been reported for use in flow cytometric analysis.Applications Tested: This R17217 antibody has been tested by flow cytometric analysis of mouse bone marrow cells. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD71 (Transferrin Receptor)
Monoclonal
0.5mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
Q62351
CD71, T9, TFR, TFR1, TR, TRFR, p90, transferrin receptor protein 1
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Murine
Flow Cytometry
R17217 (RI7 217.1.4)
FITC
Tfrc
Conjugated
Tfrc
IgG2a, kappa
50μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
22042
Videos

For Research Use Only.