Learn More
CD71 (Transferrin Receptor) Monoclonal Antibody (OKT9 (OKT-9)), PE-Cyanine7, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
£89.25 - £233.00
Specifications
Antigen | CD71 (Transferrin Receptor) |
---|---|
Clone | OKT9 (OKT-9) |
Concentration | 5 μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15508436
|
Affymetrix eBioscience
25-0719-41 |
25 Tests |
£89.25
25 tests |
Estimated Shipment: 15-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15518436
|
Affymetrix eBioscience
25-0719-42 |
100 Tests |
£233.00
100 tests |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Description: The OKT9 monoclonal antibody reacts with human CD71, a 170-180 kDa type II transmembrane protein. CD71, the transferrin receptor, exists as a homodimer on the cell surface and is essential for cellular growth. CD71 is expressed by immature proliferating cells and at low levels on resting mature lymphocytes. Antigen or mitogen stimulation of T and B cells upregulates the expression of CD71. Applications Reported: This OKT9 (OKT-9) antibody has been reported for use in flow cytometric analysis. Applications Tested: This OKT9 (OKT-9) antibody has been pre-titrated and tested by flow cytometric analysis of stimulated normal human peripheral blood cells. This can be used at 5 μL (0.5 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 μL cell sample + 100 μL IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation.
CD71 (Transferrin Receptor) Monoclonal antibody specifically detects CD71 (Transferrin Receptor) in Human samples. It is validated for Flow Cytometry.Specifications
CD71 (Transferrin Receptor) | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2 | |
2610028K12Rik; AI195355; AI426448; AU015758; CD antigen CD71; CD71; chicken transferrin receptor; E430033M20Rik; I79_000642; IMD46; mammary tumor virus receptor 1; Mtvr1; Mtvr-1; p90; sTfR; T9; TfR; tfR1; TFR2; TFRC; TR; transferrin receptor; transferrin receptor (p90, CD71); transferrin receptor 2; transferrin receptor protein 1; Transferrin receptor protein 1, serum form; Trfr | |
TFRC | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
TFRC |
OKT9 (OKT-9) | |
Flow Cytometry | |
PE-Cyanine7 | |
Mouse | |
Human | |
P02786 | |
7037 | |
IgG1 κ | |
Affinity chromatography | |
Antibody |
For Research Use Only.