missing translation for 'onlineSavingsMsg'
Learn More

CD71 (Transferrin Receptor) Monoclonal Antibody (OKT9 (OKT-9)), Functional Grade, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 16-0719-81

Code : Z2

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15238507

  • £90.00 / 50µg
Estimated Shipment: 03-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Description: The OKT9 monoclonal antibody reacts with human CD71, a 170-180 kDa type II transmembrane protein. CD71, the transferrin receptor, exists as a homodimer on the cell surface and is essential for cellular growth. CD71 is expressed by immature proliferating cells and at low levels on resting mature lymphocytes. Antigen or mitogen stimulation of T and B cells upregulates the expression of CD71. Applications Reported: This OKT9 (OKT-9) antibody has been reported for use in flow cytometric analysis. Applications Tested: This OKT9 (OKT-9) antibody has been tested by flow cytometric analysis of PHA-stimulated human peripheral blood mononuclear cells. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Storage and handling: Use in a sterile environment. Filtration: 0.2 μm post-manufacturing filtered. Purity: Greater than 90%, as determined by SDS-PAGE. Endotoxin Level: Less than 0.001 ng/μg antibody, as determined by LAL assay. Aggregation: Less than 10%, as determined by HPLC.

Cellular uptake of iron occurs via receptor-mediated endocytosis of ligand-occupied transferrin receptor into specialized endosomes. Endosomal acidification leads to iron release. The apotransferrin-receptor complex is then recycled to the cell surface with a return to neutral pH and the concomitant loss of affinity of apotransferrin for its receptor. Transferrin receptor is necessary for development of erythrocytes and the nervous system. Serum transferrin receptor (sTfR) is used as a means of detecting erythropoietin (EPO) misuse by athletes and as a diagnostic test for anemias resulting from a number of conditions including rheumatoid arthritis, pregnancy, irritable bowel syndrome and in HIV patients.
Specifications

Specifications

CD71 (Transferrin Receptor)
Monoclonal
1 mg/mL
PBS with no preservative; pH 7.2
P02786
TFRC
Affinity chromatography
RUO
7037
4° C
Liquid
Control, Flow Cytometry, Functional Assay
OKT9 (OKT-9)
Functional Grade
TFRC
2610028K12Rik; AI195355; AI426448; AU015758; CD antigen CD71; CD71; chicken transferrin receptor; E430033M20Rik; I79_000642; IMD46; mammary tumor virus receptor 1; Mtvr1; Mtvr-1; p90; sTfR; T9; TfR; tfR1; TFR2; TFRC; TR; transferrin receptor; transferrin receptor (p90, CD71); transferrin receptor 2; transferrin receptor protein 1; Transferrin receptor protein 1, serum form; Trfr
Mouse
50 μg
Primary
Human
Antibody
IgG1 κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only.