missing translation for 'onlineSavingsMsg'
Learn More

CD71 (Transferrin Receptor), Biotin, clone: OKT9 (OKT-9), eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 13-0719-80

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15289619

  • £49.99 / 25µg


Estimated Shipment date
from Supplier 03-04-2018
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: The OKT9 monoclonal antibody reacts with human CD71, a 170-180kDa type II transmembrane protein. CD71, the transferrin receptor, exists as a homodimer on the cell surface and is essential for cellular growth. CD71 is expressed by immature proliferating cells and at low levels on resting mature lymphocytes. Antigen or mitogen stimulation of T and B cells upregulates the expression of CD71.Applications Tested: This OKT9 (OKT-9) antibody has been tested by flow cytometric analysis of unstimulated and PHA-stimulated (3 days) human peripheral blood cells. This can be used at less than or equal to 0.125 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD71 (Transferrin Receptor)
OKT9 (OKT-9)
Biotin
Conjugated
Murine
25μg
Store at 2-8°C. Do not freeze.
Monoclonal
Flow Cytometry
0.5mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
TFRC
IgG1
RUO
Primary
Human
Videos