Learn More
CD7, PE, clone: CD7-6B7, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MHCD07044
Code : 50
Additional Details : Weight : 0.50000kg
Description
CD7 (gp40, Leu9) is a member of the immunoglobulin gene superfamily. Its N-terminal amino acids 1-107 are highly homologous to Ig kappa-L chains whereas the carboxy-terminal region of the extracellular domain is proline-rich and has been postulated to form a stalk from which the Ig domain projects. CD7 is expressed on the majority of immature and mature T-lymphocytes, and T cell leukemia. It is also found on natural killer cells, a small subpopulation of normal B cells and on malignant B cells. Cross-linking surface CD7 positively modulates T cell and NK cell activity as measured by calcium fluxes, expression of adhesion molecules, cytokine secretion and proliferation. CD7 associates directly with phosphoinositol 3'-kinase. CD7 ligation induces production of D-3 phosphoinositides and tyrosine phosphorylation.
R-phycoerythrin (PE) is a stable and highly soluble phycobiliprotein which provides maximal absorbance and fluorescence without susceptibility to internal or external fluorescence quenching, thus providing an exceptional quantum yields and molar extinction coefficients.Specifications
CD7 | |
Monoclonal | |
PE | |
CD7 | |
GP40, LEU-9, TP41, Tp40 | |
Mouse | |
Purified | |
ASR, ASR/IVD | |
924 | |
Store at 2 - 8 °C. | |
Liquid |
Flow Cytometry | |
CD7-6B7 | |
PBS with 4mg/mL BSA, sucrose and 0.1% sodium azide | |
P09564 | |
CD7 | |
Human CD7 | |
2 mL | |
Primary | |
Human | |
Antibody | |
IgG2a |