missing translation for 'onlineSavingsMsg'
Learn More

CD7 Mouse anti-Human, Alexa Fluor® 700, Clone: eBio124-1D1 (124-1D1), eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 56-0079-42

Code : NEW

Additional Details : Weight : 0.23750kg

Product Code. 15570327

  • £174.00 / 100 tests

In stock, ready to ship: 7

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Furthermore, it has been suggested that CD7 co-operates with CD28 during Treg function, as mice deficient in both CD28 and CD7 have reduced total numbers of Tregs and these Tregs have reduced suppressive activity. Applications Reported: This eBio124-1D1 (124-1D1) antibody has been reported for use in flow cytometric analysis. Applications Tested: This eBio124-1D1 (124-1D1) antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.5 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Alexa Fluor™ 700 emits at 723 nm and can be excited with the red laser (633 nm). Most instruments will require a 685 LP mirror and 710/20 filter. Please make sure that your instrument is capable of detecting this fluorochrome. Excitation: 633-647 nm; Emission: 723 nm; Laser: Red Laser. Filtration: 0.2 μm post-manufacturing filtered. CD7 (gp40, Leu9) is a 40 kDa member of the immunoglobulin gene superfamily. CD7 contains N-terminal amino acids 1-107 are highly homologous to Ig kappa-L chains whereas the carboxy-terminal region of the extracellular domain is proline-rich and has been postulated to form a stalk from which the Ig domain projects. CD7 is expressed on the majority of immature and mature T-lymphocytes, and T cell leukemia. Further, CD7 is also found on natural killer cells, a small subpopulation of normal B cells and on maligt B cells. Cross-linking surface CD7 positively modulates T cell and NK cell activity as measured by calcium fluxes, expression of adhesion molecules, cytokine secretion and proliferation. CD7 associates directly with phosphoinositol 3′-kinase. CD7 ligation induces production of D-3 phosphoinositides and tyrosine phosphorylation. Expression of CD7 is an important marker used in leukemia diagnostics.
Specifications

Specifications

CD7
Monoclonal
5 μL/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
P09564
GP40, LEU-9, TP41, Tp40
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
eBio124-1D1 (124-1D1)
Alexa Fluor™ 700
CD7
Liquid
CD7
IgG1, kappa
100 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
924
Videos
Documents

For Research Use Only.