Learn More
CD69 Monoclonal Antibody (FN50), PerCP-eFluor™ 710, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Affymetrix eBioscience 46-0699-42
Code : NEW
Additional Details : Weight : 0.23750kg
Description
Also called AIM or activation inducer molecule, CD69 is a gp28/34 disulfide bonded homodimer with a Mr of 60kDa under non-reducing conditions. CD69 contains one or two N linked oligosacaride. The CD69 molecule is also present on activated platelets. In normal peripheral blood a variable percentage of cells express the CD69 antigen. It is involved in lymphocyte signal transduction.
CD69 Monoclonal antibody specifically detects CD69 in Human samples. It is validated for Flow Cytometry.Specifications
CD69 | |
Monoclonal | |
5 μL/Test | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2 | |
Q07108 | |
CD69 | |
Affinity chromatography | |
RUO | |
969 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
FN50 | |
PerCP-eFluor 710 | |
CD69 | |
5830438K24Rik; Activation inducer molecule; activation inducer molecule (AIM/CD69); AI452015; AIM; BL-AC/P26; CD69; CD69 antigen; CD69 antigen (p60, early T-cell activation antigen); CD69 molecule; CLEC2C; C-type lectin domain family 2 member C; C-type lectin domain family 2, member C; EA1; early activation antigen CD69; early lymphocyte activation antigen; early T-cell activation antigen p60; GP32/28; leukocyte surface antigen Leu-23; MLR-3; VEA; Very Early Activation Antigen | |
Mouse | |
100 Tests | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
For Research Use Only.