missing translation for 'onlineSavingsMsg'
Learn More

CD69, clone: FN50, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 14-0699-80

Code : Z2

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15277057

  • £29.97 / 25µg


Estimated Shipment
28-02-2018
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: The FN50 monoclonal antibody reacts with human CD69, also known as very early activation antigen (VEA). CD69 is approximately 30kDa and is expressed on the cell-surface as a disulfide-linked dimer. CD69 is rapidly upregulated upon activation and expressed on lymphocytes, monocytes and platelets.Applications Reported: The FN50 antibody has been reported for use in flow cytometric analysis and immunohistochemical staining.Applications Tested: The FN50 antibody has been tested by flow cytometric analysis of resting and 6-hour TPA-activated human normal human peripheral blood cells. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Purity: Greater than 90%, as determined by SDS-PAGE.Aggregation: Less than 10%, as determined by HPLC.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD69
FN50
Unconjugated
Purified
Murine
25μg
Store at 2-8°C.
Monoclonal
Immunohistochemistry
0.5mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Very Early Activation Antigen, VEA
IgG1
RUO
Primary
Human
Videos