missing translation for 'onlineSavingsMsg'
Learn More

CD69, Biotin, clone: FN50, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 13-0699-80

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15249619

  • £49.99 / 25µg


Estimated Shipment date
from Supplier 26-02-2018
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: The FN50 monoclonal antibody reacts with human CD69, also known as very early activation antigen (VEA). CD69 is approximately 30kDa and is expressed on the cell-surface as a disulfide-linked dimer. CD69 is rapidly upregulated upon activation and expressed on lymphocytes, monocytes and platelets.Applications Reported: This FN50 antibody has been reported for use in flow cytometric analysis.Applications Tested: This FN50 antibody has been tested by flow cytometric analysis of resting and 6-hour TPA-activated human normal human peripheral blood cells. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD69
FN50
Biotin
Conjugated
Murine
25μg
Store at 2-8°C. Do not freeze.
Monoclonal
Flow Cytometry
0.5mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Very Early Activation Antigen, VEA
IgG1
RUO
Primary
Human
Videos