missing translation for 'onlineSavingsMsg'
Learn More

CD68, clone: eBioY1/82A (Y1/82A), eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 14-0689-80

Code : Z2

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15227057

  • £36.94 / 25Āµg


Call for latest availability
+44 (0)1509 555 500
Add to basket

Description

Description

Description: The eBioY1/82 antibody reacts with CD68 which belongs to the sialomucin family and is closely related to the family of lysosomal-associated membrane proteins (lamps) and scavenger receptor. CD68 is predominantly an intracellular protein, found mainly in the late endosomal compartment but can be detected in smaller amounts on the surface of cells of mainly myeloid derived cells; monocytes and macrophages, dendritic cells (and in some langerhan cells), neutrophils, basophils mast cells, myeloid progenitor cells, and a subset of CD34+ hemopoietic bone marrow progenitor cells. The function has not been fully elucidated but based on it's homolgy and structure suggests a role in antigen processing or presentation and protection of the lysosomal membrane from hydrolytic enzymes. Reports have also seen expression in activated T cells, and about 40% of peripheral blood B-lymphocytes and 50% of all B-ALL.Staining with eBioY1/82A shows surface staining on B cells and a minor percentage of monocytes while intracellular staining results in about 50% of B cells and most monocytes.Applications Reported: This eBioY1/82A (Y1/82A) antibody has been reported for use in flow cytometric analysis, and immunohistology staining of frozen tissue sections.Applications Tested: This eBioY1/82A (Y1/82A) antibody has been tested by intracellular staining and flow cytometric analysis. This can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Purity: Greater than 90%, as determined by SDS-PAGE.Aggregation: Less than 10%, as determined by HPLC.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD68
Monoclonal
0.5mg/mL
PBS with 0.09% sodium azide; pH 7.2
P34810
macrosialin
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow CytometryĆ¾Immunohistochemistry (Frozen)
eBioY1/82A (Y1/82A)
Unconjugated
CD68
Purified
CD68
IgG2b, kappa
25μg
Store at 2-8°C.
Primary
968
Videos

For Research Use Only.