CD68 Mouse anti-Human, Biotin, Clone: 815CU17, eBioscience™

Mouse Monoclonal Antibody

Overview
Brand: Affymetrix eBioscience

Manufacturer Part Number: 13-0687-80

25UG Anti-Human CD68 Biotin

Code: NEW

Additional Details:
Additional Details: Weight: 0.09500kg



Disclaimers: For Research Use Only.

Product Code. 15557316

Quantity Price
1 £ 49.480000000000004 / 25µg
Estimated Shipment date
from Supplier 28-03-2017
Add to basket


Description and Specification

Specification

Antigen CD68
Format Conjugated
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Host Species Mouse
Applications Immunohistochemistry (Formalin/Paraffin)
Applications Microscopy
Applications Western Blotting
Isotype IgG1
Quantity 25μg
Regulatory Status RUO
Species Reactivity Human
Storage Requirements Store at 2-8°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Clone 815CU17
Concentration 0.5mg/mL
Conjugate Biotin

This 815CU17 monoclonal antibody reacts with human CD68, a 110 kDa, transmembrane glycoprotein that belongs to the sialomucin family and is closely related to the lysosomal-associated membrane proteins (LAMP) and scavenger receptor. CD68 is predominantly an intracellular protein, found mainly in the late endosomal compartment but can be detected in smaller amounts on the surface of mainly myeloid-derived cells: monocytes, macrophages, dendritic cells (and in some langerhans cells), neutrophils, basophils, mast cells, myeloid progenitor cells, and a subset of CD34+ hematopoietic bone marrow progenitor cells. The function has not been fully elucidated but based on homology and structure, CD68 may play a role in antigen processing or presentation and protection of the lysosomal membrane from hydrolytic enzymes. Reports have also shown expression in activated T cells, and about 40% of peripheral blood B lymphocytes and 50% of all B-ALL.

This 815CU17 antibody does not cross-react with mouse or rat CD68.

For flow cytometric analysis of human CD68, we recommend clone eBioY1/82A.