Learn More
CD66b Monoclonal Antibody (G10F5), PE-Cyanine7, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Affymetrix eBioscience 25-0666-41
Code : NEW
Additional Details : Weight : 0.23750kg
Description
Description: This G10F5 monoclonal antibody reacts with human CD66b, one of four members of the CD66 family of granulocyte-specific activation antigens. Expressed on neutrophils and eosinophils, CD66b is upregulated following stimulation. This glycosyl-phosphatidylinositol-linked cell surface receptor plays a role in regulating neutrophil activation and adhesion to fibronectin, galectin-3, and E-selectin. Applications Reported: This G10F5 antibody has been reported for use in flow cytometric analysis. Applications Tested: This G10F5 antibody has been pre-titrated and tested by flow cytometric analysis of normal human perpheral blood cells. This can be used at 5 μL (0.06 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Light sensitivity: This tandem dye is sensitive photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 μL cell sample + 100 μL IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation.
CD66b Monoclonal antibody specifically detects CD66b in Human samples. It is validated for Flow Cytometry.Specifications
CD66b | |
Monoclonal | |
5 μL/Test | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2 | |
P31997 | |
CEACAM8 | |
Affinity chromatography | |
RUO | |
1088 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
G10F5 | |
PE-Cyanine7 | |
CEACAM8 | |
carcinoembryonic antigen CGM6; carcinoembryonic antigen gene family member 6; carcinoembryonic antigen related cell adhesion molecule 8; carcinoembryonic antigen-related cell adhesion molecule 8; CD66b; CD67; CD67 antigen; CEACAM8; CGM6; NCA-95; non-specific cross-reacting antigen NCA-95 | |
Mouse | |
25 Tests | |
Primary | |
Human | |
Antibody | |
IgM κ |
For Research Use Only.