missing translation for 'onlineSavingsMsg'
Learn More

CD66b, PE, clone: G10F5, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 12-0666-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15506816

  • £70.03 / 25 tests


EU Stock 1
Add to basket

Description

Description

Description: This G10F5 monoclonal antibody reacts with human CD66b, one of four members of the CD66 family of granulocyte-specific activation antigens. Expressed on neutrophils and eosinophils, CD66b is upregulated following stimulation. This glycosyl-phosphatidylinositol-linked cell surface receptor plays a role in regulating neutrophil activation and adhesion to fibronectin, galectin-3, and E-selectin.Applications Reported: This G10F5 antibody has been reported for use in flow cytometric analysis.Applications Tested: This G10F5 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.125 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD66b
Monoclonal
5μl/Test
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2
P31997
CD66b, CD67, CGM6, NCA-95
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
G10F5
PE
CEACAM8
Conjugated
CEACAM8
IgM, kappa
25 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
1088
Videos

For Research Use Only.