missing translation for 'onlineSavingsMsg'
Learn More

CD64 (Fc gamma Receptor 1), PerCP-eFluor™ 710, clone: 10.1, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 46-0649-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15529146

  • £83.31 / 25 tests


EU Stock 1
Add to basket

Description

Description

Description: The 10.1 monoclonal antibody reacts with human CD64 (FcRI), a 75kDa type I transmembrane protein. CD64 is the high affinity receptor for IgG and is involved in antibody-dependent cell-mediated cytotoxicity (ADCC), phagocytosis, and regulation of cytokine production. Monocytes and macrophages express CD64, while mature granulocytes and lymphocytes are negative.Applications Reported: This 10.1 antibody has been reported for use in flow cytometric analysis.Applications Tested: This 10.1 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. PerCP-eFluor® 710 emits at 710 nm and is excited with the blue laser (488 nm); it can be used in place of PerCP-Cyanine5.5. We recommend using a 710/50 bandpass filter, however, the 695/40 bandpass filter is an acceptable alternative. Please make sure that your instrument is capable of detecting this fluorochrome.Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 μL cell sample + 100 μL IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically.Excitation: 488 nm; Emission: 710 nm; Laser: Blue Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD64 (Fc gamma Receptor 1)
Monoclonal
5μl/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
P12314, Q92637
CD64, CD64A, FCRI, FLJ18345, IGFR1
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
10.1
PerCP-eFluor 710
FCGR1B
Conjugated
FCGR1A, FCGR1B
IgG1, kappa
25 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
2209, 2210
Videos

For Research Use Only.