missing translation for 'onlineSavingsMsg'
Learn More

CD64 (Fc gamma Receptor 1), PerCP-eFluor™ 710, clone: 10.1, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 46-0649-42

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15539146

  • £206.01 / 100 tests


EU Stock 4
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: The 10.1 monoclonal antibody reacts with human CD64 (FcRI), a 75kDa type I transmembrane protein. CD64 is the high affinity receptor for IgG and is involved in antibody-dependent cell-mediated cytotoxicity (ADCC), phagocytosis, and regulation of cytokine production. Monocytes and macrophages express CD64, while mature granulocytes and lymphocytes are negative.Applications Reported: This 10.1 antibody has been reported for use in flow cytometric analysis.Applications Tested: This 10.1 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. PerCP-eFluor® 710 emits at 710 nm and is excited with the blue laser (488 nm); it can be used in place of PerCP-Cyanine5.5. We recommend using a 710/50 bandpass filter, however, the 695/40 bandpass filter is an acceptable alternative. Please make sure that your instrument is capable of detecting this fluorochrome.Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 μL cell sample + 100 μL IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically.Excitation: 488 nm; Emission: 710 nm; Laser: Blue Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD64 (Fc gamma Receptor 1)
10.1
PerCP-eFluor 710
Conjugated
Murine
100 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Flow Cytometry
5μL (0.25μg)/test
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
FcgR1
IgG1
RUO
Primary
Human
Videos