Learn More
CD64 Monoclonal Antibody (10.1), PE, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
£79.00 - £189.00
Specifications
Clone | 10.1 |
---|---|
Concentration | 5 μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | PE |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15596806
|
Affymetrix eBioscience
12-0649-42 |
100 Tests |
£189.00
100 tests |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15586806
|
Affymetrix eBioscience
12-0649-41 |
25 Tests |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Description: The 10.1 monoclonal antibody reacts with human CD64 (FcRI), a 75 kDa type I transmembrane protein. CD64 is the high affinity receptor for IgG and is involved in antibody-dependent cell-mediated cytotoxicity (ADCC), phagocytosis, and regulation of cytokine production. Monocytes and macrophages express CD64, while mature granulocytes and lymphocytes are negative. Applications Reported: The 10.1 antibody has been reported for use in flow cytometric analysis. Applications Tested: This 10.1 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered.
CD64 (FCGR1A) encodes a protein that plays an important role in the immune response. CD64 is a high-affinity Fc-gamma receptor and is one of three related gene family members located on chromosome 1. CD64 plays an important role in the immune response and its dysfunction has been studied in cervical adenitis and tetrasomy 21.Specifications
10.1 | |
Flow Cytometry | |
PE | |
Mouse | |
Human | |
Primary | |
4° C, store in dark, DO NOT FREEZE! |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
IgG1 κ | |
Affinity chromatography | |
Antibody |
For Research Use Only.