missing translation for 'onlineSavingsMsg'
Learn More

CD64 (Fc gamma Receptor 1) Mouse anti-Human, APC-eFluor® 780, Clone: 10.1, eBioscience™

Mouse Monoclonal Antibody

£81.50 - £206.00

Specifications

Antigen CD64 (Fc gamma Receptor 1)
Clone 10.1
Target Species Human
Conjugate APC-eFluor™ 780
Isotype IgG1, kappa
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15579556
View Documents
Affymetrix eBioscience
47-0649-41
25 Tests
£81.50
25 tests
View alternative products
Save Up to 
»
Estimated Shipment
02-03-2021
Please to purchase this item. Need a web account? Register with us today!
15589556
View Documents
Affymetrix eBioscience
47-0649-42
100 Tests
£206.00
100 tests
View alternative products
Save Up to 
»
Estimated Shipment
03-02-2021
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Description: The 10.1 monoclonal antibody reacts with human CD64 (FcRI), a 75 kDa type I transmembrane protein. CD64 is the high affinity receptor for IgG and is involved in antibody-dependent cell-mediated cytotoxicity (ADCC), phagocytosis, and regulation of cytokine production. Monocytes and macrophages express CD64, while mature granulocytes and lymphocytes are negative. Applications Reported: This 10.1 antibody has been reported for use in flow cytometric analysis. Applications Tested: This 10.1 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 µL (0.125 µg) per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. APC-eFluor® 780 emits at 780 nm and is excited with the Red laser (633 nm). Please make sure that your instrument is capable of detecting this fluorochome. Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 µL of cell sample + 100 µL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat.

00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically. Excitation: 633-647 nm; Emission: 780 nm; Laser: Red Laser. Filtration: 0.2 μm post-manufacturing filtered. CD64 (FCGR1A) encodes a protein that plays an important role in the immune response. CD64 is a high-affinity Fc-gamma receptor and is one of three related gene family members located on chromosome 1. CD64 plays an important role in the immune response and its dysfunction has been studied in cervical adenitis and tetrasomy 21.
Specifications

Specifications

CD64 (Fc gamma Receptor 1)
Human
IgG1, kappa
Monoclonal
Liquid
Antibody
4° C, store in dark, DO NOT FREEZE!
10.1
APC-eFluor™ 780
Flow Cytometry
Primary
Monoclonal
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
RUO
SDS

For Research Use Only.