Learn More
CD63 Monoclonal Antibody (NVG-2), PE-Cyanine7, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
£96.75 - £246.00
Specifications
Antigen | CD63 |
---|---|
Clone | NVG-2 |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15598416
|
Affymetrix eBioscience
25-0631-82 |
100 μg |
£246.00
100µg |
Estimated Shipment: 08-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15588416
|
Affymetrix eBioscience
25-0631-80 |
25 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Description: The monoclonal antibody NVG-2 reacts with mouse CD63, also known as Lysosomal-Associated Membrane Protein 3 (LAMP-3) or tetraspanin 30 (TSPN30), a member of tetraspanin family of proteins characterized by four transmembrane domains. CD63 is expressed on a variety of cell types of hematopoietic lineage, e.g., granulocytes, B lymphocytes, platelets, as well as cells of non-hematopoietic origin. It can be found on the cell membrane, late endocytic vesicles, lysosomes, exosomes, and other specialized granules. On the cell surface, CD63 has been shown to interact with various proteins forming tetraspanin-enriched microdomains (TEM). Its high expression on the cell membrane may be indicative of cell activation, hence, CD63 is often used as an activation marker for basophils, platelets and other cells. Applications Reported: This NVG-2 antibody has been reported for use in intracellular staining followed by flow cytometric analysis. Applications Tested: This NVG-2 antibody has been tested by intracellular staining followed by flow cytometric analysis of mouse resident peritoneal exudate cells using the Intracellular Fixation & Permeabilization Buffer Set (cat. 88-8824) and protocol. Please refer to Best Protocols: Protocol A: Two step protocol for (cytoplasmic) intracellular proteins located under the Resources Tab online. This can be used at less than or equal to 0.125 μg per test.
CD63 Monoclonal antibody specifically detects CD63 in Mouse samples. It is validated for Flow Cytometry.Specifications
CD63 | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
P41731 | |
IgG2a κ | |
Affinity chromatography | |
Antibody |
NVG-2 | |
Flow Cytometry | |
PE-Cyanine7 | |
Rat | |
Mouse | |
12512 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! |
For Research Use Only.