missing translation for 'onlineSavingsMsg'
Learn More

CD63, eFluor 660, clone: H5C6, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 50-0639-42

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15559886

  • £181.37 / 100 tests


Estimated Shipment date
from Supplier 26-06-2018
Add to basket

Description

Description

Description: This H5C6 monoclonal antibody reacts with human CD63, a type III member of the tetraspanin family of transmembrane proteins. CD63 is expressed intracellularly on lysosomes, endosomes, and granules of resting platelets and basophils. However, cell surface expression of CD63 can be detected on activated basophils and platelets, monocytes, macrophages, and granulocytes. This receptor is also expressed on endothelial cells, fibroblasts, and smooth muscle cells. Studies have demonstrated that CD63 associates with integrins (VLA-3 and VLA-6) and TIMP-1 to mediate the allergic response.Applications Reported: This H5C6 antibody has been reported for use in flow cytometric analysis.Applications Tested: This H5C6 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.5 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. eFluor® 660 is a replacement for Alexa Fluor® 647. eFluor® 660 emits at 659 nm and is excited with the red laser (633 nm). Please make sure that your instrument is capable of detecting this fluorochome.Excitation: 633-647 nm; Emission: 668 nm; Laser: Red Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD63
Monoclonal
5μl/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
P08962
LAMP-3, ME491, MLA1, OMA81H, TSPAN30
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
H5C6
eFluor 660
CD63
Conjugated
CD63
IgG1, kappa
100 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
967
Videos

For Research Use Only.